Ammonium chloride-[¹⁵N] (99% ¹⁵N)
Leverandør: CAMBRIDGE ISOTOPE LABS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ammonium-15N chloride, Ammonium chloride-15N
NLM-467-5
NLM-467-50
CAMBNLM-467-5EA
0
DKK
CAMBNLM-467-5
CAMBNLM-467-50
Ammonium chloride-[¹⁵N] (99% ¹⁵N)
Ammonium chloride-[¹⁵N]
Formel:
H₄ClN |
MDL Number:
MFCD00075417 CAS-nummer: 39466-62-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...