(±)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
tert-Butyl 3-hydroxypyrrolidine-1-carboxylate, 3-Hydroxypyrrolidine, N-BOC protected
B3603-25G
B3603-5G
TCIAB3603-25GEA
1590
DKK
TCIAB3603-25G
TCIAB3603-5G
(±)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)
(±)-1-Boc-3-hydroxypyrrolidine
Formel:
C₉H₁₇NO₃ Molekylvægt: 187.24 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD04038535 CAS-nummer: 103057-44-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...