(±)-1-Boc-3-hydroxypyrrolidine ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-Butyl 3-hydroxypyrrolidine-1-carboxylate, 3-Hydroxypyrrolidine, N-BOC protected

H28750.14 H28750.06
H28750.14EA 3660 DKK
H28750.14 H28750.06
(±)-1-Boc-3-hydroxypyrrolidine ≥97%
(±)-1-Boc-3-hydroxypyrrolidine

Purity: 97% CAS No.: 103057-44-9 Molecular Formula: C9H17NO3 Formula Weight: 187.24 Melting Point: 60-62° MDL No.: MFCD04038535

Formel: C₉H₁₇NO₃
Molekylvægt: 187.24 g/mol
Smeltepunkt: 60…62 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04038535
CAS-nummer: 103057-44-9

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR