(S)-(+)-1-Boc-3-hydroxypyrrolidine 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

B3055-1G B3055-5G
TCIAB3055-1GEA 247 DKK
TCIAB3055-1G TCIAB3055-5G
(S)-(+)-1-Boc-3-hydroxypyrrolidine 98,0 GC_ASSAY_METHOD
(S)-(+)-1-Boc-3-hydroxypyrrolidine
Formel: C₉H₁₇NO₃
Molekylvægt: 187.2362 g/mol
MDL Number: MFCD01317839
CAS-nummer: 101469-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fbe974a297f4a93d45db0070be7570d9038dc75&vid=0fbe974a297f4a93d45db0070be7570d9038dc75&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C
Specific rotation [a]20/D +25.0 to +29.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR