Safety shoes, slip-on, Steelite™ MICROFIBRE FW81
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FW81WHR37 FW81WHR48 FW81WHR47 FW81WHR46 FW81WHR45 FW81WHR44 FW81WHR43 FW81WHR42 FW81WHR41 FW81WHR40 FW81WHR38 FW81WHR36 FW81WHR35 FW81BKR48 FW81BKR47 FW81BKR46 FW81BKR45 FW81BKR44 FW81BKR43 FW81BKR42 FW81BKR41 FW81BKR40 FW81BKR39 FW81WHR39 FW81BKR36 FW81BKR38 FW81BKR35 FW81BKR37 FW81BKR34 FW81WHR34 FW81WHR49
114-4426EA 402 DKK
114-4426 114-4437 114-4436 114-4435 114-4434 114-4433 114-4432 114-4431 114-4430 114-4429 114-4427 114-4425 114-4424 PRTWFW81BKR48 PRTWFW81BKR47 PRTWFW81BKR46 PRTWFW81BKR45 PRTWFW81BKR44 PRTWFW81BKR43 PRTWFW81BKR42 PRTWFW81BKR41 PRTWFW81BKR40 PRTWFW81BKR39 114-4428 PRTWFW81BKR36 PRTWFW81BKR38 PRTWFW81BKR35 PRTWFW81BKR37 PRTWFW81BKR34 PRTWFW81WHR34 PRTWFW81WHR49
Safety shoes, slip-on, Steelite™ MICROFIBRE FW81
Sko og støvler
Anti-static, slip-on lab shoes manufactured from high quality, water resistant microfibre plus a single-density lightweight PU sole. Resistant to oil.
  • Padded collars provide all day comfort
  • Shock absorbent heel
  • Machine washable at 30 °C
  • Self-cleaning sole to prevent cross-contamination
  • Protective steel toecap (200 J)
Suitable for medical and food industry.
These shoes do not utilise animal product or biproducts.
Colour: White or black.
Certificeringer: EN ISO 20345:2011; S2; AS 2210.3:2019.