Bottles, wide neck, octagonal, with standard ground stoppers
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
632414112150 632414112250 N632414112350 632414112500 632414112750 N632414212100 632414212150 632414212250 632414212350 632414212500 632414212750
KAVA632414112150EA 148 DKK
KAVA632414112150 KAVA632414112250 KAVAN632414112350 KAVA632414112500 KAVA632414112750 KAVAN632414212100 KAVA632414212150 KAVA632414212250 KAVA632414212350 KAVA632414212500 KAVA632414212750
Bottles, wide neck, octagonal, with standard ground stoppers
Flasker Glasflasker
Soda-glass, clear or amber.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
Ideal for sampling, storing and transporting powders and pastes. Amber coloured to protect light-sensitive materials.