Combined safety storage cabinets, type 90, K-PHOENIX-90 / K-PHOENIX Vol.2-90
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30333001 30334001 30335001 30336001
135-1763EA 29550 DKK
135-1763 135-1764 MSGA30335001 135-2584
Combined safety storage cabinets, type 90, K-PHOENIX-90 / K-PHOENIX Vol.2-90
Møbler Opbevaringskabinetter
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient.
Two in one: storage of flammable liquids and aggressive chemicals in one cabinet.
  • Safety elements assembled outside the storage compartment for increased protection against corrosion, easy to clean
  • Standard door open arrest system (compartment with folding doors), doors stop in any position (compartment with wing doors) - convenient and safe removal or storage of containers
  • Doors lockable with cylinder locking (integration in a locking system possible, left compartment)
  • Adjusting aids to compensate for uneven floor
  • Integrated exhaust outlet ready for connection (NW 75 mm) to a technical exhaust system with only one extraction air inlet
K-PHOENIX Vol.2-90 compartment with folding door:
Serial automatic door closing (TSA) releases the arrested door after approx. 60 seconds. Acoustic and visual signal before closing.
Serial automatic door closing (TSA) releases the arrested door after approx. 60 seconds. Acoustic and visual signal before closing.
• Furnace tested (type test) in accordance with EN 14470-1
• Fire resistance 90 minutes
• GS approval, CE conformity
• EN 14727 conformity (laboratory furniture)
Certificeringer: Furnace tested (type test) in accordance with EN 14470-1; fire resistance 90 minutes; GS approval; CE conformity; EN 14727 conformity (laboratory furniture).
Bestillingsoplysninger: Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check the website or contact Avantor for further information.
Leveringsinformation: Delivery prices available upon request.