Syringe tips, Distritips®, for DISTRIMAN® pipettes
Leverandør: GILSON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F164130 F164140 F164150 F164100 F164110 F164120
612-7818EA 814 DKK
612-7818 612-7819 GILSF164150 GILSF164100 GILSF164110 GILSF164120
Syringe tips, Distritips®, for DISTRIMAN® pipettes
Pipettespidser
Accurate PP syringes with volume level indicators. For use with the adjustable repetitive pipettor Distriman® when handling viscous, dense, volatile or hot liquids.
  • Micro 125 µl, Mini 1250 µl and Maxi 12,5 ml tips cover the entire volume range (aliquots from 1 µl to 1,25 ml)
  • More than 100 aliquots can be dispensed from each syringe
  • Free of dyes, no risk of contamination from the components
  • Sterilised tips are individually wrapped and certified free of rNase, DNase, DNA, rNA, ATP, and pyrogens