Capillary columns, TraceGOLD™ TG-OPP
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26075-5760 26076-1420 26076-2240 26075-1420
555-8818EA 7980 DKK
555-8818 555-8816 555-8814 555-8820
Capillary columns, TraceGOLD™ TG-OPP
Kromatografikolonner
Application-specific columns for organophosphorus pesticides.
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Fast analysis time giving full separation of organophosphorus pesticides
  • Ideal for US EPA methods 8141A