Graduated pipettes, Qualicolor, colour coded
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N632434026312 N632434026314 N632434026516 N632434026618 632434026719 N632434026722 N632434026723 632434026725
KAVAN632434026312EA 75.5 DKK
KAVAN632434026312 KAVAN632434026314 KAVAN632434026516 KAVAN632434026618 KAVA632434026719 KAVAN632434026722 KAVAN632434026723 KAVA632434026725
Graduated pipettes, Qualicolor, colour coded
Pipetter Pipetter med inddeling
Soda-lime glass, class B.
  • Calibrated to deliver (TD)
  • Colour coded
  • Not autoclavable at 121 °C
Certificeringer: DIN 12696, ISO 835.