Bulb pipettes, Qualicolor, colour coded (one mark), Class B
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N632433131014 N632433131016 N632433131018 N632433131019 N632433131022 N632433131023 632433131025 632433131030 632433131037
KAVAN632433131014EA 64.3 DKK
KAVAN632433131014 KAVAN632433131016 KAVAN632433131018 KAVAN632433131019 KAVAN632433131022 KAVAN632433131023 KAVA632433131025 KAVA632433131030 KAVA632433131037
Bulb pipettes, Qualicolor, colour coded (one mark), Class B
Pipetter Kolbepipetter
Soda-lime glass.
  • Enlarged form
  • Not autoclavable at 121 °C
Certificeringer: DIN 12690, ISO 648