Sleeves for ground joints, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SWN961 SWN962 SWN963 SWN964 SWN965 SWN966 SWN968 SWN969
AZLOSWN961EA 302 DKK
AZLOSWN961 AZLOSWN962 AZLOSWN963 AZLOSWN964 AZLOSWN965 AZLOSWN966 AZLOSWN968 AZLOSWN969
Sleeves for ground joints, Azlon®
Muffer med NS-slib Muffer med slib
Sleeves, PTFE, for conical ground joints.
  • Chemical resistance
  • Allows easy separation of joints
  • Intended for single or occasional use
  • Forms a gas tight seal
  • No contamination from joint grease
Working temperature: −100 to +250 °C.
Certificeringer: Suitable for food contact applications in accordance with USA regulations, USP Class VI grade PTFE.