Funnels, powder
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
632413102100 632413102060 632413102080
KAVA632413102100EA 387 DKK
KAVA632413102100 KAVA632413102060 KAVA632413102080
Funnels, powder
Tragte Pulvertragte
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
Bestillingsoplysninger: Ø = internal stem diameter.