Baths and circulators, stainless steel
Leverandør: HUBER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2038.0021.99 2057.0001.01 2056.0001.01 2058.0001.01 2059.0001.01 2060.0001.01
461-0557EA 10810 DKK
461-0557 HUBE2057.0001.01 HUBE2056.0001.01 HUBE2058.0001.01 HUBE2059.0001.01 HUBE2060.0001.01
Baths and circulators, stainless steel
Cirkulatorsystemer Åbne cirkulatorsystemer Bade med aftagelige cirkulatorer
Circulating bath thermostats with baths made from stainless steel are simple to use and suitable for many applications such as: sample temperature control, analysis, material testing or checking food. The units offer high temperature stability and are fitted with over-temperature and low level protection suitable for use with flammable fluids. A powerful pressure/suction pump ensures optimum circulation and temperature homogenity in the bath and also allows temperature control of external applications using a pump adapter (accessory). There is a range of optional accessories including inserts, variable adjustable platforms, bath covers, probes as well as tubing, temperature-control fluids and various adapters, please see www.vwr.com for details.
  • Pilot ONE® controller with plug and play technology, 14,5 cm touch screen and comfortable menu navigation
  • Precise temperature control
  • Over-temperature and low level protection
  • Powerful pressure or suction pump
  • Programmer with calendar and clock functions, allows five-point calibration
  • PT100 sensor connection, USB and LAN interfaces
The Pilot ONE® multi-touch controller features a brilliant 14,5 cm touch screen with graphic functions and easy navigation menu in 11 languages. All important operating parameters and temperature values are clearly displayed on the touch screen. The favourites menu, one-click operation and the integral technical glossary mean operation is simple. Integrated USB and ethernet ports allow connection to a PC or network, e.g. for remote control or data transmission.
MPC-controller is a modern and easy to use microprocessor controller with a large temperature display limited to essential functions only. MPC controller features over-temperature alarm and low liquid level protection. All MPC models also feature a powerful and suction pump for optimum performance.
Large temperature display
LED indicators for pump, cooling and heating
Simple operation using only three keys
Maximum pump capacities:
Pilot ONE® (CC-E): 27 L/min, 700 mbar (pressure), 25 L/min, 400 mbar (suction)
MPC: 20 L/min, 200 mbar (pressure), 17 L/min, 180 mbar (suction)
Note: The working temperature range of all models can be lowered with an optional cooling coil, connected to tap water or a chiller.
Certificeringer: Comply with DIN 12876, Safety class III, FL.
Advarsel: Note: The working temperature range of all models can be lowered with an optional cooling coil, connected to tap water or a chiller.