Cooling and heating thermostats, stainless steel, with natural refrigerant, Ministats
Leverandør: HUBER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2015.0007.01 2016.0006.01 2016.0014.04 2014.0006.01 2014.0011.01 2015.0005.01 2016.0005.01
461-0739EA 56050 DKK
461-0739 461-0740 HUBE2016.0014.04 461-1003 461-0803 461-0744 461-0745
Cooling and heating thermostats, stainless steel, with natural refrigerant, Ministats
Cirkulatorsystemer Åbne cirkulatorsystemer Termostatbade til opvarmning/køling
Their compact dimensions enable Ministats® to be placed in the smallest of spaces, for example in a laboratory fume cupboard or inside technical plant. Despite their minimal size the systems have a wide variety of features and offer performance levels suitable for temperature control of photometers, refractometers, viscometers, distillation apparatus, reaction vessels and mini-plants. They are mainly used for external applications, however the bath opening allows objects to be thermoregulated directly in a thermostat bath. A powerful pressure/suction pump with an infinitely controlled rotational speed ensures optimum circulation. The maximum pressure can also be regulated by an optional pressure sensor, meaning that sensitive glass reactors and apparatus can be protected from breakage. Ministats® have the 'professional' level Pilot ONE® controller as standard with a colour TFT display and convenient operator guidance. Analogue NAMUR connections are available via the optional Com.G@te module allowing Ministats® to be integrated into plant and process control systems. The Pilot ONE® multi-touch controller features a brilliant 14,5 cm touchscreen with graphic functions and easy navigation menu in 11 languages. All important operating parameters and temperature values are clearly displayed on the touchscreen. The favourites menu, one-click operation and the integral technical glossary mean operation is simple. Integrated USB and Ethernet ports allows connection to a PC or network, e.g. for remote control or data transmission.
  • Pilot ONE® controller with plug and play technology, 14,5 cm touchscreen and easy menu navigation
  • Precise temperature control
  • Over-temperature and low level protection
  • Powerful pressure/suction pump
  • Programmer features calendar and clock functions, allows 5-point calibration
  • PT100 sensor connection, USB and LAN interfaces
'W' denotes models are water cooled.
Max. flow rate/max pressure: 27 L/min, 700 mbar, (pressure) 20 L/min, 400 mbar (suction).
Certificeringer: Comply with DIN 12876, Safety class III, FL.
Advarsel: Cooling capacity at +20; 0; −20 and −30 °C.
With displacement insert.