HPLC columns, Purospher® STAR RP-18 endcapped
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.50036.0001 1.50037.0001 1.50225.0001 1.50231.0001 1.50239.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50253.0001 1.50254.0001 1.50255.0001 1.50256.0001 1.50358.0001 1.50359.0001 1.51455.0001 1.51456.0001 1.51460.0001 1.51448.7184 1.50596.0001 1.50615.0001 1.50620.0001 1.50414.0001 1.50617.0001 1.50470.0001 1.50431.0001 1.50469.0001 1.50471.0001 1.50622.0001 1.50427.0001 1.50468.0001 1.50626.0001 1.50398.0001 1.50413.0001 1.50627.0001 1.50607.0001 1.50621.0001 1.50593.0001 1.50625.0001 1.50624.0001 1.50612.0001 1.50428.0001 1.51914.0001 1.50037.1003 1.50251.1003 1.50252.1003 1.50358.1003 1.50359.1003 1.50431.1003 1.50617.1003 1.51455.1003 1.50036.1003 1.51456.1003 1.50393.0001
1.50036.0001EA 5850 DKK
1.50036.0001 1.50037.0001 1.50225.0001 1.50231.0001 1.50239.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50253.0001 1.50254.0001 1.50255.0001 1.50256.0001 1.50358.0001 1.50359.0001 1.51455.0001 1.51456.0001 1.51460.0001 1.51448.7184 1.50596.0001 1.50615.0001 1.50620.0001 1.50414.0001 1.50617.0001 1.50470.0001 1.50431.0001 1.50469.0001 1.50471.0001 1.50622.0001 1.50427.0001 1.50468.0001 1.50626.0001 1.50398.0001 1.50413.0001 1.50627.0001 1.50607.0001 1.50621.0001 1.50593.0001 1.50625.0001 1.50624.0001 1.50612.0001 1.50428.0001 1.51914.0001 1.50037.1003 1.50251.1003 1.50252.1003 1.50358.1003 1.50359.1003 1.50431.1003 1.50617.1003 1.51455.1003 1.50036.1003 1.51456.1003 1.50393.0001
HPLC columns, Purospher® STAR RP-18 endcapped
Kromatografikolonner
Fast method development of complex samples across the pH range from 1,5 to 10,5 with different mobile phases and temperature conditions.
Purospher® STAR RP-18 endcapped HPLC columns are designed for universal use. Basic, neutral and metal-chelating compounds can easily be separated with simple mobile phases – naturally without peak tailing. Thanks to its outstanding performance and stability, Purospher® STAR RP-18 endcapped offers maximum flexibility in method development. Robust methods can be developed over the entire pH range from 1,5 to 10,5. High pH-stability allows the separation of strongly basic compounds with alkaline eluents. The combination of high purity silica, best all-round retention characteristics, outstanding pH stability up to pH 10,5, and suitability for up to 100% aqueous mobile phases, make Purospher® STAR RP-18 endcapped an all-round top performance column, almost universal in its range of applications.
  • Highest silica purity (99,999%) for excellent peak symmetry
  • High separation efficiency
  • Absolutely reproducible results from run-to-run and batch-to-batch
  • Best all-round RP selectivity
  • No phase collapse when using highly aqueous mobile phases
  • Good suitability for LC-MS applications
These cartridges require 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a 4×4 mm guard column. LiChroCART® columns 250×10 mm require 1.51419.0001 manu-CART® 10. Short LiChroCART® columns can be ordered as a set, including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.
The Hibar® RT columns are complete with end fittings. When using a guard column with a Hibar® RT column, we recommend 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.