HPLC columns, Hypersil GOLD™ CN
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25803-012101 25802-102130 25802-052130 25805-014001 25805-154630 25805-254630 25803-103030 25803-014001 25803-011001 25802-152130 25803-154630 25805-254030
555-5713EA 4630 DKK
555-5713 555-5708 554-4320 555-5686 555-5776 554-4322 555-5729 555-5715 555-5712 555-5710 554-4321 555-5785
HPLC columns, Hypersil GOLD™ CN
Kromatografikolonner
Columns for normal and reversed phase separations.
  • Alternate selectivity with lower hydrophobicity
  • Excellent peak shape, outstanding sensitivity
  • Less retention for faster analysis