VWR® Chemical Resistant Cleanroom Overalls, Type 4/5/6
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-8231EA 5540 DKK
113-8231 113-8232 113-8233 113-8234 113-8235 113-8236
VWR® Chemical Resistant Cleanroom Overalls, Type 4/5/6
Engangsbeklædning
Disposable, anti-static overall made of a microporous film, with a nylon scrim between film and substrate, that gives the material additional strength.
  • Hood
  • Zip closure with flap
  • Elasticated cuffs, ankles and waist
  • Seams sealed with dark green tape (liquid proof)
  • Anti-static
  • Individually packed
This breathable garment provides protection against chemical dusts, paint sprays, asbestos, and other airborne particles. It can be used in any non hazardous environment where protection against dirt, grime, splashes and spills is required.
Ideal for use in Critical Environments (IEST-RP-CC003.3 – Level I). Suitable for use in ISO Class 7 + 8 cleanroom environments.
Weight: 60 g/m²
Colour: White
Certificeringer: Type 4/5/6, EN 340, EN 1149-5, EN 1073-2, EN 14126; Cat. III