Wash bottles, tracker, with coloured cap, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WTR250VT WTR500VT
215-3287EA 371 DKK
215-3287 215-3289
Wash bottles, tracker, with coloured cap, Azlon®
Flasker Sprøjteflasker
LDPE translucent.
  • Unique write-on, wipe-off panel
  • Supplied with a set of COSHH labels
  • DripLok® vapour venting cap version available for ultimate safety with volatile solvents
Wash bottle designed with complete labelling facility to ensure full traceability. Meets industry standards including those required by the clinical pathology accreditation body. Tracks set to indicate: bottle contents, dates of first use and expiry, batch number, COSHH information, prepare and date of receipt or preparation.
Certificeringer: Azlon® wash bottles meet theMore Product Information