3,5-Dibromo-4-picoline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,5-Dibromo-4-methylpyridine

D4118-5G D4118-1G
TCIAD4118-5GEA 870 DKK
TCIAD4118-5G TCIAD4118-1G
3,5-Dibromo-4-picoline ≥98.0% (by GC, titration analysis)
3,5-Dibromo-4-picoline
Formel: C₆H₅Br₂N
Molekylvægt: 250.92 g/mol
MDL Number: MFCD00128859
CAS-nummer: 3430-23-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7a0d9cdbb243421b9784efbd3aaf07a4f8bd095&vid=b7a0d9cdbb243421b9784efbd3aaf07a4f8bd095&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR