Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: N-Ethyl-1-((4-(phenyldiazenyl)phenyl)diazenyl)naphthalen-2-amine
O0096-25G O0096-5G
TCIAO0096-25GEA 887 DKK
TCIAO0096-25G TCIAO0096-5G
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Formel: C₂₄H₂₁N₅
Molekylvægt: 379.4570 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003904
CAS-nummer: 6368-72-5
EINECS: 228-862-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=885b4f06dc02af334a8138b58a27a4b2de8fba98&vid=885b4f06dc02af334a8138b58a27a4b2de8fba98&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Orange to Brown to Dark red powder to crystal
Lambda max. 531.0 to 535.0 nm (MeOH)
Absorbance (E1%1cm) min. 750 (MeOH, 531.0 to 535.0 nm)