Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Sodium isethionate, Sodium hydroxyethyl sulphonate, 2-Hydroxyethanesulphonic acid sodium salt

H0241-25G H0241-500G
TCIAH0241-25GEA 197 DKK
TCIAH0241-25G TCIAH0241-500G
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) ≥98.0% (by titrimetric analysis)
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate)
Formel: C₂H₅NaO₄S
Molekylvægt: 148.11 g/mol
MDL Number: MFCD00007534
CAS-nummer: 1562-00-1
EINECS: 216-343-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 191.0 to 196.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR