1-Benzhydrylpiperazin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3760-5G B3760-25G
TCIAB3760-5GEA 485 DKK
TCIAB3760-5G TCIAB3760-25G
1-Benzhydrylpiperazin ≥98.0% (by GC, titration analysis)
1-Benzhydrylpiperazin
Formel: C₁₇H₂₀N₂
Kogepunkt: 185°C
MDL Number: MFCD00038379
CAS-nummer: 841-77-0
EINECS: 212-667-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d3f1703d57f7f0f5d12c175924e1c65ae34eaee&vid=2d3f1703d57f7f0f5d12c175924e1c65ae34eaee&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR