7-Methylquinolin ≥75.0% (by GC) (contains 25% 5-form at maximum)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M0417-25G M0417-5G
TCIAM0417-25GEA 567 DKK
TCIAM0417-25G TCIAM0417-5G
7-Methylquinolin ≥75.0% (by GC) (contains 25% 5-form at maximum)
7-Methylquinolin
Formel: C₁₀H₉N
Molekylvægt: 143.19 g/mol
MDL Number: MFCD00006805
CAS-nummer: 612-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf947e39e4c79459f274702eecebcda09a09d9a6&vid=1b6fe562c2993e6d390441ab8070a52addd4aadd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 75.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR