4-Brom-2-methyl-6-nitroanilin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene
B3730-1G
B3730-5G
TCIAB3730-1GEA
132
DKK
TCIAB3730-1G
TCIAB3730-5G
4-Brom-2-methyl-6-nitroanilin ≥98.0% (by GC)
4-Brom-2-methyl-6-nitroanilin
Formel:
C₇H₇BrN₂O₂ Molekylvægt: 231.05 g/mol Smeltepunkt: 155 |
MDL Number:
MFCD00052919 CAS-nummer: 77811-44-0 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...