5-Azaindol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine

P1896-1G P1896-100MG
TCIAP1896-1GEA 1520 DKK
TCIAP1896-1G TCIAP1896-100MG
5-Azaindol ≥98.0% (by GC, titration analysis)
5-Azaindol
Formel: C₇H₆N₂
Molekylvægt: 118.14 g/mol
Kogepunkt: 132°C
MDL Number: MFCD00955936
CAS-nummer: 271-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e98350e3fa4b1e30d66f49ab366764a95e314d4c&vid=e98350e3fa4b1e30d66f49ab366764a95e314d4c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR