3-(4-Bromphenyl)propionsyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B3753-5G
B3753-1G
TCIAB3753-5GEA
329
DKK
TCIAB3753-5G
TCIAB3753-1G
3-(4-Bromphenyl)propionsyre ≥98.0% (by GC, titration analysis)
3-(4-Bromphenyl)propionsyre
Formel:
C₉H₉BrO₂ Kogepunkt: 250°C |
MDL Number:
MFCD01310793 CAS-nummer: 1643-30-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...