1-Chlor-4-(1-(4-chlorphenyl)-2,2-dichlorethyl)benzol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,2-Bis(4-chlorophenyl)-1,1-dichloroethane, 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane, p,p'-TDE, 4,4'-DDD, p,p'-DDD, TDE

B0132-5G B0132-1G
TCIAB0132-5GEA 534 DKK
TCIAB0132-5G TCIAB0132-1G
1-Chlor-4-(1-(4-chlorphenyl)-2,2-dichlorethyl)benzol ≥98.0% (by GC)
4,4'-TDE
Formel: C₁₄H₁₀Cl₄
Kogepunkt: 406 °C (1013 hPa)
Smeltepunkt: 111 °C
Massefylde: 1,476 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000851
CAS-nummer: 72-54-8
EINECS: 200-783-0
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,03061
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1ed63e1edc7a595f8d6704d363c3499c10e104&vid=8a1ed63e1edc7a595f8d6704d363c3499c10e104&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 109.0 to 112.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR