Chrysen ≥98.0% (by GC), purified by sublimation

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Chrysene, Benzo[a]phenanthrene, 1,2-Benzophenanthrene

C0339-1G C0339-100MG
TCIAC0339-1GEA 2830 DKK
TCIAC0339-1G TCIAC0339-100MG
Chrysen ≥98.0% (by GC), purified by sublimation
Chrysene
Formel: C₁₈H₁₂
Kogepunkt: 448°C
Smeltepunkt: 250…252 °C
Massefylde: 1,274 g/cm³ (20 °C)
MDL Number: MFCD00003698
CAS-nummer: 218-01-9
EINECS: 205-923-4
UN nummer: 3077
ADR: 9,III
Merck Index: 14,02255
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=818364151cf7b86c1226d381ac23966dac7d0a81&vid=1acbbe5a6795729d1809a48c26dc64a98079f069&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR