3-Chlor-4-methoxybenzoesyre ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Chloro-p-anisic acid

C2550-25G C2550-5G
TCIAC2550-25GEA 2890 DKK
TCIAC2550-25G TCIAC2550-5G
3-Chlor-4-methoxybenzoesyre ≥97.0% (by GC, titration analysis)
3-Chlor-4-methoxybenzoesyre
Formel: C₈H₇ClO₃
Molekylvægt: 186.59 g/mol
MDL Number: MFCD00016512
CAS-nummer: 37908-96-6
EINECS: 253-708-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19295e4b1b73c9fb99f07b14f4dbf87e43a15f99&vid=19295e4b1b73c9fb99f07b14f4dbf87e43a15f99&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 215.0 to 219.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR