Thymol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol
M0410-25G
M0410-500G
TCIAM0410-25GEA
132
DKK
TCIAM0410-25G
TCIAM0410-500G
Thymol ≥99.0% (by GC)
Thymol
Formel:
2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH Molekylvægt: 150.22 g/mol Kogepunkt: 232°C Flammepunkt: 104 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002309 CAS-nummer: 89-83-8 EINECS: 201-944-8 UN nummer: 1759 ADR: 8 III Merck Index: 14,09399 |
|
Specifikationer Testresultater
Appearance | White powder to lump |
Purity (GC) | min. 99.0 % |
Melting point | 49.0 to 52.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...