Dysprosium(III) carbonate tetrahydrate ≥99.9%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
15286.09
15286.18
15286.09EA
742
DKK
15286.09
15286.18
Dysprosium(III) carbonate tetrahydrate ≥99.9%
Dysprosium(III) carbonate tetrahydrate
Purity: 99.9%
CAS No.: 38245-35-1
Molecular Formula: Dy2(CO3)3.4H2O
Formula Weight: 577.06 (505.03anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00150229
Formel:
C₃Dy₂O₉·4H₂O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00150229 CAS-nummer: 38245-35-1 EINECS: 225-770-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...