3,3'-Dimethyldiphenylamine 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Di-m-tolylamine

B21765.03 B21765.06
B21765.03EA 668 DKK
B21765.03 B21765.06
3,3'-Dimethyldiphenylamine 98%
3,3'-Dimethyldiphenylamine

Purity: 98% CAS No.: 626-13-1 Molecular Formula: C14H15N Formula Weight: 197.28 Flash Point: 110°(230°F) Refractive Index: 1.6225 Boiling Point: 200°/24mm MDL No.: MFCD00059315

Formel: C₁₄H₁₅N
Molekylvægt: 197.28 g/mol
Kogepunkt: 200 °C (24 mmHg)
Massefylde: 1.04
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00059315
CAS-nummer: 626-13-1

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR