Hexanophenone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H0116-25G H0116-500G
TCIAH0116-25GEA 444 DKK
TCIAH0116-25G TCIAH0116-500G
Hexanophenone ≥98.0% (by GC)
Hexanophenone
Formel: C₁₂H₁₆O
MDL Number: MFCD00009512
CAS-nummer: 942-92-7
EINECS: 213-394-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8ee47ac1eb856897d0d3aaf3c5499d00ec7ea57&vid=d8ee47ac1eb856897d0d3aaf3c5499d00ec7ea57&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR