(3-Carboxypropyl)triphenylphosphonium bromide ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A19302.14
A19302.22
A19302.36
A19302.14EA
801
DKK
A19302.14
A19302.22
A19302.36
(3-Carboxypropyl)triphenylphosphonium bromide ≥97%
(3-Carboxypropyl)triphenylphosphonium bromide
Purity: 97%
CAS No.: 17857-14-6
Molecular Formula: HO2C(CH2)3P(C6H5)3Br
Formula Weight: 429.30
Melting Point: 244-250°
Beilstein Reference: 4173599
Formel:
C₂₂H₂₂BrO₂P Smeltepunkt: 244…250 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00274196 CAS-nummer: 17857-14-6 |