3,4'-Dinitrobenzophenone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D2551-25G D2551-5G
TCIAD2551-25GEA 1460 DKK
TCIAD2551-25G TCIAD2551-5G
3,4'-Dinitrobenzophenone ≥98.0% (by GC)
3,4'-Dinitrobenzophenone
Formel: C₁₃H₈N₂O₅
MDL Number: MFCD00617174
CAS-nummer: 1469-74-5
UN nummer: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b7449aedd743655aa352c474206902feb010448&vid=7b7449aedd743655aa352c474206902feb010448&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in hot Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR