2-Hydroxy-4-octyloxybenzophenone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2-Hydroxy-4-(octyloxy)phenyl)(phenyl)methanone

H0288-25G H0288-500G
TCIAH0288-25GEA 132 DKK
TCIAH0288-25G TCIAH0288-500G
2-Hydroxy-4-octyloxybenzophenone ≥98.0% (by GC)
2-Hydroxy-4-octyloxybenzophenone
Formel: C₂₁H₂₆O₃
Molekylvægt: 326.44 g/mol
MDL Number: MFCD00027327
CAS-nummer: 1843-05-6
EINECS: 217-421-2
Merck Index: 14,06742
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cb6ae93748cd1b6b0050513623931755a5f52ea&vid=7cb6ae93748cd1b6b0050513623931755a5f52ea&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR