Geranyl acetate ≥70% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(E)-3,7-Dimethylocta-2,6-dien-1-yl acetate
G0028-25ML
G0028-500ML
G0028-100ML
TCIAG0028-25MLEA
165
DKK
TCIAG0028-25ML
TCIAG0028-500ML
TCIAG0028-100ML
Geranyl acetate ≥70% (by GC)
Geranyl acetate
Formel:
C₁₂H₂₀O₂ Molekylvægt: 196.29 g/mol Kogepunkt: 120°C Flammepunkt: 118°C |
MDL Number:
MFCD00015037 CAS-nummer: 105-87-3 EINECS: 203-341-5 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 70.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...