3-Hydroxyisonicotinic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Hydroxypyridine-4-carboxylic acid
H1377-1G
H1377-5G
TCIAH1377-1GEA
600
DKK
TCIAH1377-1G
TCIAH1377-5G
3-Hydroxyisonicotinic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxyisonicotinic acid
Formel:
C₆H₅NO₃ Smeltepunkt: 315°C |
MDL Number:
MFCD00234165 CAS-nummer: 10128-71-9 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...