2,2,2-Trifluoroethyl formate ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T2586-5G
T2586-1G
TCIAT2586-5GEA
673
DKK
TCIAT2586-5G
TCIAT2586-1G
2,2,2-Trifluoroethyl formate ≥95.0% (by GC)
2,2,2-Trifluoroethyl formate
Formel:
C₃H₃F₃O₂ Kogepunkt: 60°C Flammepunkt: 7°C |
MDL Number:
MFCD06797733 CAS-nummer: 32042-38-9 UN nummer: 3272 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...