3',4'-Dimethylbenzophenone-2-carboxylic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D2978-5G D2978-1G
TCIAD2978-5GEA 1060 DKK
TCIAD2978-5G TCIAD2978-1G
3',4'-Dimethylbenzophenone-2-carboxylic acid ≥98.0% (by titrimetric analysis)
3',4'-Dimethylbenzophenone-2-carboxylic acid
Formel: C₁₆H₁₄O₃
CAS-nummer: 2159-42-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a33f7751467a80a215aed3b3ff736185f57cebc&vid=7a33f7751467a80a215aed3b3ff736185f57cebc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR