1-Methoxy-2-butanol ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M0747-25ML M0747-500ML
TCIAM0747-25MLEA 140 DKK
TCIAM0747-25ML TCIAM0747-500ML
1-Methoxy-2-butanol ≥93.0% (by GC)
1-Methoxy-2-butanol
Formel: C₅H₁₂O₂
Molekylvægt: 104.15 g/mol
Kogepunkt: 135°C
Flammepunkt: 39°C
MDL Number: MFCD00059143
CAS-nummer: 53778-73-7
EINECS: 258-763-2
UN nummer: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af5763519925269cd6e7276d3610dfac2fccdcad&vid=af5763519925269cd6e7276d3610dfac2fccdcad&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR