2'-Hydroxy-3-phenylpropiophenone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H1313-25G
H1313-5G
TCIAH1313-25GEA
1060
DKK
TCIAH1313-25G
TCIAH1313-5G
2'-Hydroxy-3-phenylpropiophenone ≥98.0% (by GC)
2'-Hydroxy-3-phenylpropiophenone
Formel:
C₁₅H₁₄O₂ Kogepunkt: 158°C |
MDL Number:
MFCD00002221 CAS-nummer: 3516-95-8 EINECS: 222-521-4 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...