4'-tert-Butylpropiophenone ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B1609-1ML B1609-5ML
TCIAB1609-1MLEA 452 DKK
TCIAB1609-1ML TCIAB1609-5ML
4'-tert-Butylpropiophenone ≥95.0% (by GC)
4'-tert-Butylpropiophenone
Formel: C₁₃H₁₈O
Kogepunkt: 162°C
MDL Number: MFCD00100434
CAS-nummer: 71209-71-7
EINECS: 275-265-0
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=487a52e59ae2985b7bb31819f339b7cd936ce801&vid=487a52e59ae2985b7bb31819f339b7cd936ce801&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR