Benzyl-4-bromophenyl ketone ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1-(4-Bromophenyl)-2-phenylethanone
B2020-1G
B2020-5G
TCIAB2020-1GEA
321
DKK
TCIAB2020-1G
TCIAB2020-5G
Benzyl-4-bromophenyl ketone ≥97.0% (by GC)
Benzyl-4-bromophenyl ketone
Formel:
C₁₄H₁₁BrO Molekylvægt: 275.14 g/mol |
MDL Number:
MFCD00016331 CAS-nummer: 2001-29-8 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 111.0 to 115.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...