Phenyl-2-pyridyl ketone oxime ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Phenyl 2-Pyridyl Ketoxime
P0223-1G
P0223-5G
TCIAP0223-1GEA
156
DKK
TCIAP0223-1G
TCIAP0223-5G
Phenyl-2-pyridyl ketone oxime ≥98.0% (by titrimetric analysis)
Phenyl-2-pyridyl ketone oxime
Formel:
C₁₂H₁₀N₂O Molekylvægt: 198.22 g/mol |
MDL Number:
MFCD00006310 CAS-nummer: 1826-28-4 EINECS: 217-374-8 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...