Benzophenone dimethyl acetal ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Benzophenone dimethyl ketal, Dimethoxydiphenylmethane

B1999-25G B1999-5G
TCIAB1999-25GEA 977 DKK
TCIAB1999-25G TCIAB1999-5G
Benzophenone dimethyl acetal ≥98.0% (by GC)
Benzophenone dimethyl acetal
Formel: C₁₅H₁₆O₂
Molekylvægt: 228.2863 g/mol
Kogepunkt: 290°C
MDL Number: MFCD00025900
CAS-nummer: 2235-01-0
EINECS: 218-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f47e7137b6e55085c3980635ee769e464547a462&vid=f47e7137b6e55085c3980635ee769e464547a462&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR