Methyl-2-benzoylbenzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B1231-25G B1231-500G
TCIAB1231-25GEA 165 DKK
TCIAB1231-25G TCIAB1231-500G
Methyl-2-benzoylbenzoate ≥98.0% (by GC)
Methyl-2-benzoylbenzoate
Formel: C₁₅H₁₂O₃
Kogepunkt: 352°C
MDL Number: MFCD00017187
CAS-nummer: 606-28-0
EINECS: 210-112-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8cebdde2dc4f7900b7eca9abfeef7386ee90878&vid=e8cebdde2dc4f7900b7eca9abfeef7386ee90878&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 53.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR