2-Chloro-5-nitrobenzophenone ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(2-Chloro-5-nitrophenyl)(phenyl)methanone
H35800.14
H35800.22
H35800.14EA
726
DKK
H35800.14
H35800.22
2-Chloro-5-nitrobenzophenone ≥98%
2-Chloro-5-nitrobenzophenone
Formel:
C₁₃H₈ClNO₃ Smeltepunkt: 78…84 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007295 CAS-nummer: 34052-37-4 EINECS: 251-811-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...