2-Amino-5-methylbenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Aminotoluene-3-sulphonic acid

A0328-25G A0328-500G
TCIAA0328-25GEA 132 DKK
TCIAA0328-25G TCIAA0328-500G
2-Amino-5-methylbenzenesulphonic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-5-methylbenzenesulphonic acid
Formel: C₇H₉NO₃S
Molekylvægt: 187.22 g/mol
MDL Number: MFCD00007908
CAS-nummer: 88-44-8
EINECS: 201-831-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1d69b790e50bbb77d4217b852e9520ec08677c8&vid=a1d69b790e50bbb77d4217b852e9520ec08677c8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR